Learn More
Streck Reagents for VC-100 Acid-Base Titration Pipet Verification Instrument
Acid-base titration method that verifies the volume delivered by pipets and automatic dilutors
Manufacturer:  Streck 270025
 View more versions of this product
Catalog No. 11-716-211
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Verifying the volume delivered by pipets and dilutors
1 Bottle titration solution, titration cartridge, 3 vials each A and B reference control, 3 vials titration acid, 50 titration cups, pipet labels
VC-100 Acid base titration pipet verification instrument
Pipet verification kit