Streck Reagents for VC-100 Acid-Base Titration Pipet Verification Instrument
Acid-base titration method that verifies the volume delivered by pipets and automatic dilutors
Manufacturer: Streck 270050
 View more versions of this product
Catalog No. 11-716-212
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With VC-100 Acid base titration pipet verification instrument
    Applications Verifying the volume delivered by pipets and dilutors
    Includes 2 Bottles titration solution, 2 titration cartridges, 3 vials each A and reference control, 5 vials titration acid, 50 titration cups, pipet labels
    Type Pipet verification kit
  • Description & Specifications
Related Products
Product Content Feedback