Learn More
Streck Retic-Chex™ II Stains and Controls: Stains Available on GSA/VA Contract for Federal Government customers only.
For counting red blood cell reticulocytes
Manufacturer:  Streck 285410
 View more versions of this product
Catalog No. 23-046568
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Retic-Chex™ II Methylene blue stain
Optimal staining of Reticulocytes for both patient and control preparations
CE
2 x 15mL
1 Year
Beckman Coulter™ STKS™, MAXM™, HmX, LH 500, Abbott CELL-DYN™ 3500 and 3700
2° to 10°C