Learn More
Streck Retic-Chex™ Linearity Assays Available on GSA/VA Contract for Federal Government customers only.
Verify the accuracy and linear reportable range of hematology instruments by comparing the laboratory's reportable range with these specialized assays
Manufacturer:  Streck 285532
 View more versions of this product
Catalog No. 23-046-626
  Check Availability
Add to cart
Disclaimers:
U.S. Patent 5,432,089.
Specifications Description & Specifications
Specifications
Retic-Chex™ linearity assay
105 Days
Siemens medical solutions diagnostics ADVIA™ 120/2120, Sysmex™ XE-2100™, Sysmex™ XE-5000™, Sysmex™ XT-2000i™
5 x 2.5mL
Vial
To establish the patient reportable range and linearity of the system
18° to 30°C