Learn More
Streck STaK-Chex™ Controls for Beckman Coulter™ Instrument Available on GSA/VA Contract for Federal Government customers only.
A complete assay for the five-part white cell differential
Manufacturer:  Streck 225503
 View more versions of this product
Catalog No. 11-716-337
Add to cart
This item is not returnable. View return policy
Description
Description
  • STaK-Chex is a whole blood five-part white cell differential control formulated for the Beckman Coulter STKS™, MAXM™, HmX and LH500·S™
  • STaK-Chex is manufactured at clinically recognized normal, low and high abnormal levels for all parameters
  • The whole blood control offers distinct populations of lymphocytes, monocytes, neutrophils, eosinophils and basophils
  • White blood cell percentages vary for each cell type and are within clinically significant ranges
  • The precision and accuracy of the white cell differential is controlled each time STaK-Chex is analyzed
  • STaK-Chex assay value files are available for download for the LH 500 and HmX. Assay sheets are bar coded and can be scanned with the Coulter wand
  • STaK-Chex has a 105-day closed-vial and a 14-day open-vial stability
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Order Info
Packaged in 4.0mL, pierceable-cap vials
Specifications
Specifications
STaK-Chex™ Control
Vial
Beckman Coulter™ STKS™, MAXM™, HmX and LH 500
3 x 4mL
1 Low, 1 Normal, 1 High
105 Days
2° to 10°C
For evaluating the accuracy and precision of automated and semi automated procedures that measure Blood cell parameters