Streck STaK-Chex™ Controls for Beckman Coulter™ Instrument Available on GSA/VA Contract for Federal Government customers only.

A complete assay for the five-part white cell differential

Manufacturer: Streck 225512

 View more versions of this product

Catalog No. 23-285-533

  Check Availability
Add to Cart

    Specifications

    Product Type STaK-Chex™ control
    Format Vial
    Stability 105 Days
    Includes Assay disc
    For Use With (Application) For evaluating the accuracy and precision of automated and semi automated procedures that measure Blood cell parameters
    For Use With (Equipment) Beckman Coulter™ STKS™, MAXM™, HmX and LH 500
    Storage Requirements 2° to 10°C
    Quantity 12 x 4mL,12 x 4mL
    Level 4 Low, 4 Normal, 4 High,4 Low, 4 Normal, 4 High

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit