Learn More
Streck Temp-Chex ™ Thermometers Available on GSA/VA Contract for Federal Government customers only.
Includes a wall chart for recording temperatures, initials and supervisor approval
Manufacturer:  Streck 240053
 View more versions of this product
Catalog No. 11-716-187D
Add to cart
Description
Description
·    Enviro-Safe: Utilizes a non-toxic, nonflammable biodegradable liquid and dye. Available for freezer, refrigerator, incubators and room temperatures.
·    Red Spirit: Mercury-free. Monitors temperatures in sub-low freezers and freezers, refrigerators, incubators and room temperatures.
·    Contained in a propylene glycol vial
·    Enclosed in a plastic tube to protect thermometer and to contain contents in case of breakage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Certifications
NIST traceable/DKD traceable· Each thermometer engraved with serial number traceable to NIST/DKD
Specifications
Specifications
0.5°
50°C
NIST, DKD
Temperature wall chart
±0.5°C
20° to 50°C
20°C
Incubator
Enviro-Safe™ Liquid