Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Strep II Tag Antibody, Alexa Fluor™ 647, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP241076AF647
Description
Strep II Tag Polyclonal antibody specifically detects Strep II Tag in Epitope Tag samples. It is validated for Western Blot, ELISASpecifications
Strep II Tag | |
Polyclonal | |
Western Blot, ELISA | |
fibronectin type III, immunoglobulin and leucine rich repeat domains 1, FIGLER1, leucine rich repeat and fibronectin type III domain containing 3, leucine-rich repeat and fibronectin type-III domain-containing protein 3, MGC2656, SALM4, synaptic adhesion-like molecule 4, UNQ5865/PRO34192 | |
Antibody Strep II Tag antibody was raised against the amino acid sequence "asWSHPQFEKga". | |
0.1 mL | |
Epitope Tags | |
Human, Mouse, Rat | |
Purified | |
Epitope Tag |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction