Learn More
IBA US DISTRIBUTION CENTER Strep-Tactin Superflow 50% suspension, 20 ml DFS Item
Manufacturer:  IBA US DISTRIBUTION CENTER 21206010
Strep-Tactin Superflow is used for purification of recombinant Strep-tag proteins under gravity flow and low pressure or FPLC . Bulk resin is recommended for packing of our columns and cartridges since the resin is optimized for column affinity chromatography rather than for batch purification.Binding capacity: 1 ml sedimented resin (corresponding to 2 ml of a 50 % suspension) is useful for the one-step purification of 50 to 100 nmol recombinant Strep-tag protein (up to 3 mg in case of a 30 KDa protein).!!!Test our new Strep-TactinXT Superflow resin!!!
Catalog No. NC0095598