Learn More
IBA LifeSciences Strep-TactinXT Superflow high capacity 50% suspension, 4 ml  DFS Item
Manufacturer:  IBA LifeSciences 24030002
Strep-TactinXT Superflow high capacity is used for purification of recombinant Strep-tagII and Twin-Strep-tag fusion proteins under gravity flow and low pressure or FPLC. The Strep-TactinXT Superflow high capacity resin provides 2 to 3 times higher binding capacity for Strep-tag proteins than standard Strep-TactinXT Superflow and therefore, higher yields of highly pure Strep-tag protein.Dynamic binding capacity: 15mg protein/ml resin, determined with mCherry-Twin-Strep-tag (30kDa).
Catalog No. NC1481822