Learn More
IBA LifeSciences Strep-tagII peptide; 1.8 mg
Supplier:  IBA LifeSciences 21018002
Encompass_Preferred
Synthetic Strep-tagII peptide recognized by Strep-Tactin, Strep-TactinXT as well as the monoclonal antibodies, StrepMAB-Classic and StrepMAB-Immo. It can be used for competitive elution of Strep-tagII fusion proteins or as control protein in dot blot assays.
Catalog No. 50-210-0131
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.