Learn More
Seracare Life Sciences IncSupplier Diversity Partner STREPTAVIDIN DYLIGHT 680  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52700025
Streptavidin is a 60,000 Dalton protein isolated from the bacterium Streptomyces avidinii. Electrophoretically pure streptavidin was labeled with DyLight 680 using a proprietary method. DyLight conjugates provide increased brightness and greater protostability than other fluorophores (ex. - FITC). The conjugate is designed for use in immunofluorescense assays including flow cytometry, immunohistology, Western blotting, microarrays and FLISA.
Catalog No. 50-671-50