Learn More
GE Healthcare Streptavidin HP SpinTrap Buffer Kit Available on GSA/VA Contract for Federal Government customers only.
Optimized for standard affinity-based enrichment/immunoprecipitation. GE Healthcare Streptavidin HP SpinTrap Buffer Kit includes high-quality, pre-made buffers to facilitate fast, reproducible protein enrichment. Sufficient for performing 16 reactions.
Manufacturer:  GE Healthcare 28913568
Catalog No. 45-002-411
Add to cart
Description
Description
Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation according to the protocol provided with Streptavidin HP SpinTrap columns.
  • High quality buffers: 2 binding/washing, 1 elution and 1 blocking buffer
  • All buffer components listed and defined; instructions included
  • Convenient format—stock solutions can be diluted directly into the provided bottles for storage
  • Eliminates time-consuming buffer preparation, which allows fast, reproducible protein enrichment
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
2 x 5mL Binding/washing buffer (TBS) (containing 0.5 M Tris and 1.5 M NaCl,pH 7.5)
3mL Elution buffer (containing 1 M Glycine-HCl, pH 2.9)
20mL Blocking buffer (containing 2 mM Biotin in TBS pH 7.5)
Instructions
1 Piece
4 to 30°C