Learn More
Novus Biologicals™ Streptavidin - HRP Antibody Labeling Kit
Streptavidin is purified from bacterium Streptomyces avidinii, is tetrameric protein that is composed of identical subunits, with tetramer having molecular weight of approximately 53kda
$179.00 - $329.00
Specifications
Product Type Streptavidin - HRP Antibody Labeling Kit
Storage Requirements Store at -20° C.
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
20-101-000 View Documents Novus Biologicals™
20101000
0.1mg Each for $329.00
Add to cart
20-100-100 View Documents Novus Biologicals™
20100100
1mg Each for $179.00
Add to cart
Description
Description
  • Tetrameric protein that is composed of identical subunits, with tetramer having molecular weight of approximately 53 kDa
Specifications
Specifications
Streptavidin - HRP Antibody Labeling Kit
Store at -20° C.
For research use only.