Learn More
BD Streptavidin, PE-CF594
Developed as a better alternative to PE-Texas Red, for use in Flow Cytometry
$158.00 - $362.00
Specifications
Concentration | 0.2 mg/mL |
---|---|
Conjugate | PE-CF594 |
Content And Storage | 4°C |
For Use With (Application) | Flow Cytometry |
Description
The Streptavidin is conjugated to BD Horizon PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496 nm and 564 nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).
Specifications
0.2 mg/mL | |
PE-CF594 | |
4°C | |
Flow Cytometry |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.