Learn More
Invitrogen™ eBioscience™ Streptavidin PE-eFluor™ 610 Non-distribution product as customer accommodation.
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
Manufacturer:  Invitrogen™ 61431780
 View more versions of this product
Catalog No. 50-112-4813
Add to cart
Alert:
Contains Sodium azide
Specifications
Specifications
0.2mg/mL
Flow Cytometric Analysis
2° to 8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling & experimental procedures.
SA, Sav
PE-eFluor 610
25μg
Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer