Learn More
Seracare Life Sciences IncSupplier Diversity Partner STREPTAVIDIN RPE-LABELED  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52700021
Streptavidin labeled with R-Phycoerythrin (RPE). Streptavidin is a protein isolated from the bacterium Streptomyces avidinii and has no carbohydrate moieties. It has a molecular weight of approximately 60,000 Daltons with an isoelectric point of 5. Product is in liquid form.
Catalog No. 50-670-35