Learn More
Molecular Probes™ Streptavidin, Unconjugated Non-distribution product as customer accommodation.
Streptavidin, Unconjugated
Manufacturer:  Molecular Probes™ SNN1001
Catalog No. SNN1001
Add to cart
Description
Description
Purified from Streptomyces avidinii and provided in lyophilized form. The streptavidin/avidin-biotin staining and detection strategy is used when increased assay sensitivity is desired for a wide variety of systems.
Format/Formulation: Unconjugated
Specifications
Specifications
Unconjugated
Solid, Lyophilized powder
Streptavidin
1mg
1mg
2 to 8°C
1 Each
Streptavidin, 1mg, Solid, Lyophilized Powder Form, Unconjugated, 0.22μm Filtration, Used When Increased Assay Sensitivity is Desired for a Wide Variety of Systems, 2 to 8°C Storage Temperature, Wet Ice Shipping Condition
Solid, lyophilized powder
Streptavidin
For laboratory use
1mg
1mg
PBS
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Laboratory Use