Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus streptokinase Antibody, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NB100644030.1ML
Description
Streptococcus streptokinase Polyclonal specifically detects Streptococcus streptokinase in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Streptococcus streptokinase | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.4) with 0.09% Sodium Azide | |
Purified Streptococcus streptokinase | |
0.1 mL | |
Primary | |
Bacteria | |
Purified |
Functional Assay, ELISA, Western Blot | |
5.0 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Functional | |
Sheep | |
Protein G purified | |
RUO | |
NB100-64403 recognises intact streptokinase, a metallo-enzyme produced by various strains of Streptococcus that can liquify blood clots by converting plasminogen to plasmin. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction