Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streptococcus streptokinase Sheep anti-Bacteria, Polyclonal, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 1 publication
$176.85 - $436.00
Specifications
Antigen | Streptococcus streptokinase |
---|---|
Concentration | 5.0 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Functional |
Applications | Western Blot, ELISA, Functional Assay |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10064403
|
Novus Biologicals
NB10064403 |
0.5 mL |
Each for $436.00
|
|
NB017575
|
Novus Biologicals
NB100644030.1ML |
0.1 mL |
Each for $176.85
|
|
Description
Streptococcus streptokinase Polyclonal specifically detects Streptococcus streptokinase in Bacteria samples. It is validated for Western Blot, ELISA.Specifications
Streptococcus streptokinase | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Functional | |
Polyclonal | |
Purified | |
RUO | |
PBS (pH 7.4) with 0.09% Sodium Azide | |
IgG | |
Protein G purified | |
NB100-64403 recognises intact streptokinase, a metallo-enzyme produced by various strains of Streptococcus that can liquify blood clots by converting plasminogen to plasmin. |
5.0 mg/mL | |
Western Blot, ELISA, Functional Assay | |
Unconjugated | |
Sheep | |
Bacteria | |
Purified Streptococcus streptokinase | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title