Learn More
Streptococcuspneumoniae Mouse anti-Bacteria, Janelia Fluor 646, Clone: 128/390, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NB10064569JF646
Catalog No. NB064569646
Add to cart
Description
Description
Streptococcuspneumoniae Monoclonal antibody specifically detects Streptococcuspneumoniae in Bacteria samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Streptococcuspneumoniae
128/390
Immunocytochemistry/Immunofluorescence
Purified
Mouse
IgG1
0.1mL
Primary
In a simple ELISA using Biogenesis Test Groups (BTGs) this is broadly reactive with S.pneumoniae. A small cross reactivity is observed with Haemophilus influenzae. Immunohistology: Reactive with isolates fixed by phenol, formalin and boiling.
Immunocytochemistry, Immunofluorescence
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
Pneumococcal surface protein A, PSPA
Native, NCTC 10319
Protein A purified
Store at 4°C in the dark.
Monoclonal
Bacteria