Streptozocin, Tocris

DNA alkylator; antitumor and induces diabetes

Manufacturer: R&D Systems 1621 /100

Quantity 100mg

 View more versions of this product

Catalog No. 16-21-1-00

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Solid

    Specifications

    Formula Weight 265.22
    Quantity 100mg
    Physical Form Solid
    CAS 18883-66-4
    Synonym 2-Deoxy-2-[[(methylnitrosoamino)carbonyl]amino]-D-glucose
    Assay Percent Range >95%
    Molecular Formula C8H15N3O7
    Recommended Storage Store at -20°C
    Chemical Name or Material Streptozocin

    Store at -20°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit