Learn More
AdipoGen Streptozotocin,Chemical. CAS: 18883-66-4
SDP
Supplier:  AdipoGen AGCN20046M050
Encompass_Preferred
Streptozotocin, AdipoGen Life Sciences. Chemical. CAS: 18883-66-4. Formula: C8H15N3O7. MW: 265.2. Antibiotic. Diabetogenic. Diabetes inducer. Induces diabetes mellitus in animal models through its toxic effects on pancreatic beta-cells. Mutagenic. Potent alkylating agent. Potent DNA methylating agent. Nitric oxide (NO) donor. Vasorelaxant. Cytotoxic to cells that express GLUT2 glucose transporter. O-GlcNAc-selective N-acetyl-beta-D-glucosaminidase (O-GlcNAcase) inhibitor. Genotoxic. Induces DNA damage. Produces DNA strand breaks. Cell death inducer. Antineoplastic. Anti-cancer agent used in chemotherapy. Induces cell cylce arrest at G2.
Catalog No. 50-464-382
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.