Learn More
GSC Go Science Crazy Stringless Pendulum Products for Science Education
Apparatus shows harmonic motion with no strings attached. Allows students to see the period of an oscillating body from different perspective
Manufacturer:  GSC Go Science Crazy PENDM01
Catalog No. S28118
Add to cart
Alert:
Not a toy. Product intended for ages 13 and up.
Description
Description
  • Shows simple harmonic motion
  • No strings needed like a traditional pendulum
  • Includes 2 each 19mm steel balls
Specifications
Specifications
Shows harmonic motion without use of strings
Stringless pendulum
L x W x H: 500 x 20 x 100mm; 850g
2 each 19mm steel balls