Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Striped Warning Tapes
Indicates hazardous areas clearly, marks physical hazards
Supplier: Brady™ 55297
Description
- Permanent adhesive stands up to tough indoor conditions
- Colors are sealed beneath protective layer of abrasion resistant clear vinyl
- Holds on to almost any clean, dry surface
Specifications
Red, White | |
Rubber-based Pressure Sensitive Adhesive | |
54 ft. | |
3 in. |
7.62 cm | |
Striped Warning Tape | |
16.4 m | |
B-950 Vinyl |
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction