Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Stripette™ Individually Paper-Plastic Wrapped Disposable Serological Pipets, Cleanroom Pack Available on GSA/VA Contract for Federal Government customers only.

Packaged in “clean room” technique for a safety and convenience revolution. Corning™ Stripette™ Individually Paper-Plastic Wrapped Disposable Serological Pipets, Cleanroom Pack is ideal for cell-based vaccine and drug production facilities or other laboratory clean rooms.

Manufacturer:  Corning™ 7017

 View more versions of this product

Catalog No. 07-200-739


Add to cart

Description

Description

Ideal for cell culture and other applications that require aseptic liquid handling. Sterile pipets feature color-coded stripes for easy volume reading and quick size identification.

  • Bidirectional graduations allow the choice of using ascending or descending scales
  • Negative graduations on the 1, 2, 5, 10, 25, and 50mL Stripettes provide additional working volume
  • 100mL Stripettes allow you to perform large-volume fluid transfers safely and easily; eliminate the need for graduated cylinders and other liquid-handling vessels
  • Cylindrical anti-drip tip design on 25, 50, and 100mL sizes reduces accidental drippage and assures accurate delivery
  • 10mL size has a wide tip for handling viscous fluids
  • Nonpyrogenic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

±2%
1.69 oz., 50mL
Purple
Yes
14.56 in.
10mL
Round
50mL
1.69 oz.
Non-pyrogenic, DNase, RNase-free
Yes
0.5mL
37cm
Tripled bagged
Stripette™ Serological Pipet
PS (Polystyrene)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit