Corning™ Stripettor™ Ultra Pipet Controller Accessory, Hydrophobic PTFE Filters

Extreme chemical resistance of PTFE membranes makes them very useful for filtering solvents or other aggressive chemicals for which other membranes are unsuitable.

Manufacturer: Corning™ 4992

 View more versions of this product

Catalog No. 07-202-352

  Check Availability
Add to Cart

Alert

Because of their hydrophobicity, PTFE membranes must be prewetted with a solvent, such as ethanol, before aqueous solutions can be filtered.


    Specifications

    For Use With (Equipment) Stripettor™ Ultra Pipet Controller
    Quantity Pack of 5
    Filter Type Polytetrafluorethylene (PTFE)
    Pore Size 0.45μm
    Color White
    Product Type Hydrophobic PTFE Filter

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit