Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRN4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548441

Catalog No. PA548441

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Binds calmodulin in a calcium dependent manner. May function as scaffolding or signaling protein.
Specifications

Specifications

STRN4
Polyclonal
PBS with 0.09% sodium azide
Q9NRL3
Striatin-4, Zinedin, STRN4, ZIN
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29888
Western Blot
Unconjugated
STRN4
Liquid
STRN4
KLH conjugated synthetic peptide between 124-153 amino acids from the N-terminal region of human STRN4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit