Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Strong Iodine Solution, 5% (w/v), USP, Spectrum™ Chemical
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Description
Spectrum™ Chemical Strong Iodine Solution, 5 Percent (w/v) (DEA List I Chemical), USP is used primarily as an antiseptic and disinfectant in pharmaceutical applications. All Spectrum Chemical USP products, including all USP iodine products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 0.05, 0.95 |
Packaging | Amber Glass Bottle |
Quantity | 4 x 4 L |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction