Alfa Aesar™ Strontium 2,4-pentanedionate hydrate

Manufacturer: Alfa Aesar™ 1461414

Quantity 25g

 View more versions of this product

Catalog No. AA1461414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12193-47-4
    Formula Weight 285.84 (Anhydrous)
    MDL Number MFCD00013502
    Synonym Strontium acetylacetonate
    Chemical Name or Material Strontium 2,4-pentanedionate hydrate

    Specifications

    Formula Weight 285.84 (Anhydrous)
    Physical Form Powder
    MDL Number MFCD00013502
    CAS 12193-47-4
    Synonym Strontium acetylacetonate
    Solubility Information Soluble in warm methoxyethanol, pyridine
    Melting Point 200°C (dec.)
    Quantity 25g
    Chemical Name or Material Strontium 2,4-pentanedionate hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit