Alfa Aesar™ Strontium 2-ethylhexanoate, 40% in 2-ethylhexanoic acid (8-12% Sr)

Manufacturer: Alfa Aesar™ 4071906

Quantity 5g

 View more versions of this product

Catalog No. AA4071906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2457-02-5
    Formula Weight 374.04
    MDL Number MFCD00070438
    Synonym 2-Ethylhexanoic acid strontium salt
    Chemical Name or Material Strontium 2-ethylhexanoate

    Specifications

    Formula Weight 374.04
    Health Hazard 3 GHS P Statement
    P280h
    Wear protective gloves/clothing.
    Physical Form Liquid
    CAS 2457-02-5
    Synonym 2-Ethylhexanoic acid strontium salt
    Health Hazard 2 GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.
    MDL Number MFCD00070438
    TSCA TSCA
    Health Hazard 1 Warning
    Odor Mild
    Concentration 40% in 2-ethylhexanoic acid (8 to 12% Sr)
    Quantity 5g
    Chemical Name or Material Strontium 2-ethylhexanoate

    GHS H Statement
    H361
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P280h
    Wear protective gloves/clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit