Strontium bromide hexahydrate, 95%, Alfa Aesar™

Manufacturer: Alfa Aesar 7210536

Quantity 500g

 View more versions of this product

Catalog No. AA7210536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7789-53-9
    Physical Form Crystalline
    Assay Percent Range 95%
    Molecular Formula Br2Sr·6H2O
    Formula Weight 355.53; 247.44 (anhydrous)
    MDL Number MFCD00150361
    Chemical Name or Material Strontium bromide hexahydrate

    Specifications

    Formula Weight 355.53; 247.44 (anhydrous)
    Quantity 500g
    EINECS Number 233-969-5
    Physical Form Crystalline
    Assay Percent Range 95%
    Molecular Formula Br2Sr·6H2O
    MDL Number MFCD00150361
    CAS 7789-53-9
    Chemical Name or Material Strontium bromide hexahydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit