Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium Chloride, 6-Hydrate, Low in Magnesium, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle
10025-70-4
SrCl2·6H2O
10025-70-4
SrCl2·6H2O

High quality chemicals for laboratory and specialized industrial use

Formula Weight 266.62
Physical Form Crystals
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.96kg
Quantity 125g
Chemical Name or Material Strontium Chloride, 6-Hydrate, Low in Magnesium