Learn More
Strontium chloride, anhydrous, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Strontium chloride is widely used in fireworks for the source of red color. It is also used in dental and biological research. It is the precursor material to other compounds of strontium. It is used to prepare yellow strontium chromate, a corrosion inhibitor for aluminum. It is also employed in glass-making, sea water aquaria, and metallurgy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | 1250°C |
Melting Point | 873°C |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.5%, metal basis |
Solubility Information | Freely soluble in water. Very slightly soluble in ethanol and acetone. |
Formula Weight | 158.53 |
Refractive Index | 1.65 |
Sensitivity | Hygroscopic |
Density | 3.052 |
Show More |
Safety and Handling
Hazard Category | H318-H335 |
Hazard Statement | In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. |
Precautionary Statement | P261-P271-P280-P304+P340-P305+P351+P338-P310-P312-P501c |
EINECSNumber | 233-971-6 |
RTECSNumber | WK8400000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.