Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium chloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
10476-85-4
SrCl2
This item is not returnable. View return policy
10476-85-4
SrCl2

Calcium sensing receptor (CaSR) agonist

Calcium sensing receptor (CaSR) agonist. Activates ERK1/2 signaling, intracellular calcium mobilization, and calcitonin secretion. Enhances osteogenic differentiation of mesenchymal stem cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Formula Weight 158.53
Percent Purity >98%
Quantity 50mg
Chemical Name or Material Strontium chloride

Recommended Storage : Desiccate at Room Temperature