Alfa Aesar™ Strontium cyclohexanebutyrate, AAS

Manufacturer: Alfa Aesar™ 3973503

Quantity 1g

 View more versions of this product

Catalog No. AA3973503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62638-05-5
    Formula Weight 426.11
    MDL Number MFCD00050727
    Synonym Cyclohexanebutyric acid strontium salt; Strontium 4-cyclohexylburate
    Chemical Name or Material Strontium cyclohexanebutyrate, AAS

    Specifications

    Formula Weight 426.11
    MDL Number MFCD00050727
    Synonym Cyclohexanebutyric acid strontium salt; Strontium 4-cyclohexylburate
    TSCA TSCA
    Physical Form Powder
    CAS 62638-05-5
    Quantity 1g
    Chemical Name or Material Strontium cyclohexanebutyrate, AAS

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit