Alfa Aesar™ Strontium dodecairon nonadecaoxide, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 2256914

Quantity 25g

 View more versions of this product

Catalog No. AA2256914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12023-91-5
    Formula Weight 1061.77
    MDL Number MFCD00075632
    Synonym Strontium ferrite
    Chemical Name or Material Strontium dodecairon nonadecaoxide

    Specifications

    Formula Weight 1061.77
    Melting Point >450°C
    TSCA TSCA
    Mesh Size −325 Mesh
    Physical Form Powder
    MDL Number MFCD00075632
    Synonym Strontium ferrite
    Quantity 25g
    CAS 12023-91-5
    Chemical Name or Material Strontium dodecairon nonadecaoxide
    Assay 99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit