Strontium Fluoride, 99.99% (Metal Basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 10878-02

Quantity 0.5g

 View more versions of this product

Catalog No. AA1087802

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 7783-48-4
    Physical Form Powder
    Assay Percent Range 99.99%, metal basis
    Molecular Formula SrF2
    Formula Weight 125.62
    MDL Number MFCD00011251
    Chemical Name or Material Strontium Fluoride

    Specifications

    Formula Weight 125.62
    Quantity 0.5g
    EINECS Number 232-000-3
    Physical Form Powder
    Assay Percent Range 99.99%, metal basis
    Molecular Formula SrF2
    MDL Number MFCD00011251
    CAS 7783-48-4
    Health Hazard 1 Irritating to eyes, respiratory system and skin.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.
    Chemical Name or Material Strontium Fluoride

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

    Irritating to eyes, respiratory system and skin.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit