Alfa Aesar™ Strontium hexafluoro-2,4-pentanedionate

Manufacturer: Alfa Aesar™ 1470506

Quantity 5g

 View more versions of this product

Catalog No. AA1470506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 121012-89-3
    Formula Weight 501.75
    MDL Number MFCD00064760
    Synonym Strontium hexafluoroacetylacetonate
    Chemical Name or Material Strontium hexafluoro-2,4-pentanedionate

    Specifications

    Formula Weight 501.75
    Physical Form Crystalline
    MDL Number MFCD00064760
    Melting Point ≈260°C (dec.)
    Quantity 5g
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    CAS 121012-89-3
    Synonym Strontium hexafluoroacetylacetonate
    Boiling Point 220°C (0.02mmHg) (subl.)
    Color White
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Strontium hexafluoro-2,4-pentanedionate

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit