Strontium hydroxide octahydrate, Technical grade, Alfa Aesar™

Manufacturer: Alfa Aesar 1219936

Quantity 500g

 View more versions of this product

Catalog No. AA1219936

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1311-10-0
    Physical Form Crystalline
    Molecular Formula Sr(OH)2·8H2O
    Formula Weight 265.76 (121.60 Anhydrous)
    MDL Number MFCD00150395
    Chemical Name or Material Strontium hydroxide octahydrate
    Grade Technical Grade

    Specifications

    Formula Weight 265.76 (121.60 Anhydrous)
    Quantity 500g
    EINECS Number 242-367-1
    Physical Form Crystalline
    Density 1.9g/mL
    Molecular Formula Sr(OH)2·8H2O
    MDL Number MFCD00150395
    CAS 1311-10-0
    Chemical Name or Material Strontium hydroxide octahydrate
    Grade Technical Grade

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit