Learn More
Strontium iodide, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Strontium iodide is used as a scintillation gamma radiation detector. It is also used in a range of hand-held radiation detection instruments as well as in medical, industrial and environmental applications. It is used in medicine as a substitute for potassium iodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 515°C |
Color | White to Yellow |
Physical Form | Powder |
Quantity | 25 g |
Merck Index | 14,8844 |
Solubility Information | Soluble in water and ethanol. |
Formula Weight | 341.43 |
Percent Purity | 99.99% |
Odor | Odorless |
Sensitivity | Hygroscopic; Light sensitive |
Show More |
Safety and Handling
Hazard Category | H290-H315-H319-H335 |
Hazard Statement | GHS H Statement H290-H335-H315-H319 May be corrosive to metals. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P234-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P390-P501c |
EINECSNumber | 233-972-1 |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.