Alfa Aesar™ Strontium iodide, anhydrous, 99% (metals basis)

Manufacturer: Alfa Aesar™ 4726822

Quantity 100g

 View more versions of this product

Catalog No. AA4726822

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 10476-86-5
    Formula Weight 341.43
    MDL Number MFCD00049557
    Merck Index 14,8844
    Chemical Name or Material Strontium iodide, Anhydrous

    Specifications

    Formula Weight 341.43
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00049557
    Health Hazard 2 GHS H Statement
    H290-H335-H315-H319
    May be corrosive to metals.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    Odor Odorless
    CAS 10476-86-5
    Merck Index 14,8844
    Melting Point 515°C
    Health Hazard 1 Warning
    Physical Form Powder
    Sensitivity Light Sensitive & Hygroscopic
    Density 4.550g/mL
    TSCA TSCA
    Color Pale yellow to dark brown
    Quantity 100g
    Chemical Name or Material Strontium iodide, Anhydrous
    Assay 97%, (metals basis)

    GHS H Statement
    H290-H335-H315-H319
    May be corrosive to metals.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit