Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Strontium neodecanoate, Sr 16-21%

Manufacturer:  Alfa Aesar™ 3957409

Quantity 10g

 View more versions of this product

Catalog No. AA3957409


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

106705-37-7
430.1
MFCD00070439
Strontium neodecanoate

Specifications

Sr 16 to 21%
106705-37-7
430.1
Strontium neodecanoate
10g
Powder
MFCD00070439
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit