Strontium nitrate, 99.999% (metals basis), Alfa Aesar™, Puratronic™

Manufacturer: Alfa Aesar 4319322

Quantity 100g

 View more versions of this product

Catalog No. AA4319322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10042-76-9
    Physical Form Crystalline, Packaged Under Argon
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula Sr(NO3)2
    Formula Weight 211.63
    MDL Number MFCD00011248
    Chemical Name or Material Strontium nitrate

    Specifications

    Physical Form Crystalline, Packaged Under Argon
    Formula Weight 211.63
    Quantity 100g
    EINECS Number 233-131-9
    Assay Percent Range 99.999% (metals basis)
    Molecular Formula Sr(NO3)2
    MDL Number MFCD00011248
    CAS 10042-76-9
    Chemical Name or Material Strontium nitrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit