Learn More
Strontium nitrate, ACS, 99.0% min, Thermo Scientific Chemicals
Description
Strontium nitrate is used in the production of road flares and in fireworks. It serves as a colorant (red) in pyrotechnics and is also used as an oxidizer in pyrotechnics. Mixed with glycolic acid, it is used to reduce the sensation of skin irritation. It is also used in electrophysiology experiments.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 570°C |
Physical Form | Crystalline |
Quantity | 1 kg |
Assay Percent Range | 99.0% min. |
Solubility Information | Soluble in water and ammonia. Slightly soluble in alcohol,ethanol and acetone. Insoluble in nitric acid. |
IUPAC Name | strontium(2+) dinitrate |
Formula Weight | 211.63 |
Grade | ACS Reagent |
Sensitivity | Hygroscopic |
Density | 2.99 g/mL |
Show More |
Safety and Handling
Hazard Category | H272-H318 |
Precautionary Statement | P210-P220-P221-P280-P305+P351+P338-P310-P370+P378q-P501c |
DOTInformation | Hazard Class: 5.1; Packaging Group: III |
EINECSNumber | 233-131-9 |
RTECSNumber | WK9800000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.