Alfa Aesar™ Strontium nitrate, anhydrous, 99.97% (metals basis)

Manufacturer: Alfa Aesar™ 3048614

Quantity 25g

 View more versions of this product

Catalog No. AA3048614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10042-76-9
    Linear Formula Sr(NO3)2
    Formula Weight 211.63
    MDL Number MFCD00011248
    UN Number UN1507
    Merck Index 14,8845
    Chemical Name or Material Strontium nitrate, Anhydrous

    Specifications

    Formula Weight 211.63
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011248
    Quantity 25g
    Density 2.990g/mL
    RTECS Number WK9800000
    Physical Form Crystalline
    Linear Formula Sr(NO3)2
    UN Number UN1507
    Color White
    Melting Point 570°C
    Health Hazard 1 Danger
    Odor Odorless
    Sensitivity Hygroscopic
    CAS 10042-76-9
    Merck Index 14,8845
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: STRONTIUM NITRATE
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H272-H302-H315-H319-H335
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material Strontium nitrate, Anhydrous
    Assay 99.97%, (metals basis)

    GHS H Statement
    H272-H302-H315-H319-H335
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit