Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strontium oxalate, 95%, Thermo Scientific Chemicals

Catalog Number p-7050840
Click to view available options
Quantity:
500 g
814-95-9
C2O4Sr
175.638
MFCD00050803
KQAGKTURZUKUCH-UHFFFAOYSA-L
strontium oxalate, unii-7iaw7e69ni, 7iaw7e69ni, strontium oxalate, src2o4, ethanedioic acid, strontium salt 1:1, oxalic acid, monostrontium salt, strontium 2+ ion oxalate, oxalic acid, strontium salt 1:1 8ci
69947
strontium;oxalate
C(=O)(C(=O)[O-])[O-].[Sr+2]
814-95-9
C2O4Sr
175.638
MFCD00050803
KQAGKTURZUKUCH-UHFFFAOYSA-L
strontium oxalate, unii-7iaw7e69ni, 7iaw7e69ni, strontium oxalate, src2o4, ethanedioic acid, strontium salt 1:1, oxalic acid, monostrontium salt, strontium 2+ ion oxalate, oxalic acid, strontium salt 1:1 8ci
69947
strontium;oxalate
C(=O)(C(=O)[O-])[O-].[Sr+2]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 500 g
UN Number UN3288
Solubility Information Soluble in hydrochloric acid and nitric acid
Formula Weight 175.64
Percent Purity 95%
Physical Form Powder or Lumps
Chemical Name or Material Strontium oxalate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 212-415-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit