Alfa Aesar™ Strontium oxalate, 95%

Manufacturer: Alfa Aesar™ 3957322

Quantity 100g

 View more versions of this product

Catalog No. AA3957322

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 814-95-9
    Formula Weight 175.64
    MDL Number MFCD00050803
    UN Number UN3288
    Chemical Name or Material Strontium oxalate

    Specifications

    Physical Form Powder/Lump
    Formula Weight 175.64
    Health Hazard 3 GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00050803
    CAS 814-95-9
    Solubility Information Soluble in hydrochloric acid and nitric acid
    Health Hazard 1 Warning
    TSCA TSCA
    UN Number UN3288
    Health Hazard 2 GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
    Quantity 100g
    Chemical Name or Material Strontium oxalate
    Assay 95%

    GHS H Statement
    H302-H312
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P280-P301+P312-P312-P363-P322-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Wash contaminated clothing before reuse.
    Specific measures (see label).
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit