Strontium perchlorate trihydrate, 98%, Alfa Aesar™

Manufacturer: Alfa Aesar 4557736

Quantity 500g

 View more versions of this product

Catalog No. AA4557736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15650-09-6
    Physical Form Crystalline
    Assay Percent Range 98%
    Molecular Formula Cl2O8Sr·3H2O
    Formula Weight 340.54 (286.52 Anhydrous)
    MDL Number MFCD09039117
    Chemical Name or Material Strontium perchlorate trihydrate

    Specifications

    Formula Weight 340.54 (286.52 Anhydrous)
    Quantity 500g
    Physical Form Crystalline
    Assay Percent Range 98%
    Molecular Formula Cl2O8Sr·3H2O
    MDL Number MFCD09039117
    CAS 15650-09-6
    Chemical Name or Material Strontium perchlorate trihydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit