Learn More
Strontium peroxide, 12.3% available oxygen, Thermo Scientific Chemicals
Description
Applications of Strontium Peroxide include pyrotechnics i.e. fireworks (red color), ammunitions (tracer/hunting), oxidizer, alarm rocket. It is also used in textiles for fibers bleaching and as an oxidizing agent for hair bleaching (professional).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 215°C (decomposition) |
Color | Brown |
Physical Form | Powder |
Quantity | 1 kg |
Linear Formula | SrO2 |
UN Number | UN1509 |
Merck Index | 14,8847 |
Solubility Information | Insoluble in water and slowly decomposes in water. |
Formula Weight | 119.62 |
Sensitivity | Moisture sensitive |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H272-H318-H335-H315 May intensify fire oxidizer. Causes serious eye damage. May cause respiratory irritation. Causes skin irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
DOTInformation | Hazard Class: 5.1 |
EINECSNumber | 215-224-6 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.